4-Bromo-3-iodophenol 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR16668-1G
OR16668-250MG
APOSOR16668-1GEA
82.8
EUR
APOSOR16668-1G
APOSOR16668-250MG
4-Bromo-3-iodophenol 95%
4-Bromo-3-iodophenol
Formel:
C₆H₄BrIO Molecular Weight: 298,904 g/mol Schmelzpunkt: 83…84 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00070745 CAS-Nummer: 202865-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...