3-Bromo-2-methylbenzaldehyde 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-6-formyltoluene, 3-Bromo-o-tolualdehyde
OR16673-1G
OR16673-250MG
OR16673-5G
APOSOR16673-1GEA
24.8
EUR
APOSOR16673-1G
APOSOR16673-250MG
APOSOR16673-5G
3-Bromo-2-methylbenzaldehyde 97%
3-Bromo-2-methylbenzaldehyde
Formel:
C₈H₇BrO Molecular Weight: 199,045 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11847533 CAS-Nummer: 83647-40-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...