5-Iodo-1H-benzimidazole 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Iodo-1H-benzo[d]imidazole
OR16685-1G
OR16685-250MG
OR16685-5G
APOSOR16685-1GEA
79.9
EUR
APOSOR16685-1G
APOSOR16685-250MG
APOSOR16685-5G
5-Iodo-1H-benzimidazole 97%
5-Iodo-1H-benzimidazole
Formel:
C₇H₅IN₂ Schmelzpunkt: 144…145 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659631 CAS-Nummer: 78597-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...