2-Bromothiophene-3-carboxaldehyde 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromothiophene-3-carbaldehyde, 2-Bromo-3-formylthiophene, 2-Bromo-3-thenaldehyde

OR16845-1G OR16845-5G OR16845-250MG
APOSOR16845-1GEA 29 EUR
APOSOR16845-1G APOSOR16845-5G APOSOR16845-250MG
2-Bromothiophene-3-carboxaldehyde 97%
2-Bromothiophene-3-carboxaldehyde
Formel: C₅H₃BrOS
Molecular Weight: 191.046 g/mol
Siedepunkt: 60 °C (0.3 mm Hg)
Lagertemperatur: Kühlschrank
MDL: MFCD09025880
CAS-Nummer: 1860-99-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR