Diethyl 3,4-dihydroxythiophene-2,5-dicarboxylate

Lieferant: Apollo Scientific

Synonyme: 2,5-Bis(ethoxycarbonyl)thiophene-3,4-diol

OR16953-5G OR16953-25G
APOSOR16953-5GEA 72.5 EUR
APOSOR16953-5G APOSOR16953-25G
Diethyl 3,4-dihydroxythiophene-2,5-dicarboxylate
Diethyl 3,4-dihydroxythiophene-2,5-dicarboxylate
Formel: C₁₀H₁₂O₆S
Molecular Weight: 260,264 g/mol
Schmelzpunkt: 132…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094035
CAS-Nummer: 1822-66-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR