Methyl 5-bromothiophene-3-carboxylate 99%

Lieferant: Apollo Scientific
Achtung

OR17258-1G OR17258-5G
APOSOR17258-1GEA 24.8 EUR
APOSOR17258-1G APOSOR17258-5G
Methyl 5-bromothiophene-3-carboxylate 99%
Methyl 5-bromothiophene-3-carboxylate
Formel: C₆H₅BrO₂S
Molecular Weight: 221,072 g/mol
Schmelzpunkt: 38…43 °C
Lagertemperatur: Kühlschrank
MDL: MFCD13386488
CAS-Nummer: 88770-19-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR