4-Amino-3,5-dichloropyridazine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Dichloropyridazin-4-amine

OR17303-100MG OR17303-250MG OR17303-1G
APOSOR17303-100MGEA 52.2 EUR
APOSOR17303-100MG APOSOR17303-250MG APOSOR17303-1G
4-Amino-3,5-dichloropyridazine
4-Amino-3,5-dichloropyridazine
Formel: C₄H₃Cl₂N₃
Molecular Weight: 163.993 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01647251
CAS-Nummer: 53180-76-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR