7-(Aminomethyl)quinoline

Lieferant: Apollo Scientific

Synonyme: (Quinolin-7-yl)methylamine

OR17563-100MG OR17563-1G OR17563-250MG
APOSOR17563-100MGEA 690 EUR
APOSOR17563-100MG APOSOR17563-1G APOSOR17563-250MG
7-(Aminomethyl)quinoline
7-(Aminomethyl)quinoline
Formel: C₁₀H₁₀N₂
Molecular Weight: 158,2 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12031213
CAS-Nummer: 773092-54-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR