2-Bromo-6-methyl-4-nitrophenol 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromo-2-hydroxy-5-nitrotoluene, 6-Bromo-4-nitro-o-cresol
OR17670-1G
OR17670-5G
OR17670-250MG
APOSOR17670-1GEA
105
EUR
APOSOR17670-1G
APOSOR17670-5G
APOSOR17670-250MG
2-Bromo-6-methyl-4-nitrophenol 95%
2-Bromo-6-methyl-4-nitrophenol
Formel:
C₇H₆BrNO₃ Schmelzpunkt: 118…122 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD26392591 CAS-Nummer: 4186-49-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...