2-Bromo-5-iodophenyl methyl sulphone 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-4-iodo-2-(methylsulphonyl)benzene
OR17675-1G
OR17675-5G
OR17675-250MG
APOSOR17675-1GEA
174
EUR
APOSOR17675-1G
APOSOR17675-5G
APOSOR17675-250MG
2-Bromo-5-iodophenyl methyl sulphone 95%
2-Bromo-5-iodophenyl methyl sulphone
Formel:
C₇H₆BrIO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD28155004 CAS-Nummer: 1820619-59-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...