3-Bromo-4-chlorobenzeneacetic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-4-chlorophenylacetic acid, 2-(3-Bromo-4-chlorophenyl)acetic acid

OR17683-1G OR17683-5G OR17683-25G
APOSOR17683-1GEA 21.9 EUR
APOSOR17683-1G APOSOR17683-5G APOSOR17683-25G
3-Bromo-4-chlorobenzeneacetic acid 95%
3-Bromo-4-chlorobenzeneacetic acid
Formel: C₈H₆BrClO₂
Molecular Weight: 249,489 g/mol
Schmelzpunkt: 95…97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD18390582
CAS-Nummer: 90004-80-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR