5,6-Dihydrouridine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)dihydropyrimidine-2,4(1H,3H)-dione, 1-[(2S,3S,4R,5S)-3,4-Dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,3-diazinane-2,4-dione

OR17694-1G OR17694-250MG OR17694-5G
APOSOR17694-1GEA 217 EUR
APOSOR17694-1G APOSOR17694-250MG APOSOR17694-5G
5,6-Dihydrouridine 95%
5,6-Dihydrouridine
Formel: C₉H₁₄N₂O₆
Molecular Weight: 246,217 g/mol
Schmelzpunkt: 101…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00047490
CAS-Nummer: 5627-05-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR