2-Nitrophenylacetylene 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Ethynyl-2-nitrobenzene
OR17873-1G
OR17873-250MG
OR17873-5G
APOSOR17873-1GEA
56.7
EUR
APOSOR17873-1G
APOSOR17873-250MG
APOSOR17873-5G
2-Nitrophenylacetylene 98%
2-Nitrophenylacetylene
Formel:
C₈H₅NO₂ Molecular Weight: 147,131 g/mol Schmelzpunkt: 82…85 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01113917 CAS-Nummer: 16433-96-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...