4-{[(2-Amino-4-hydroxypteridin-6-yl)methyl]amino}benzoic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Pteroic acid, 4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzoic acid, Folic Acid Impurity D
OR17919-1G
OR17919-5G
OR17919-250MG
APOSOR17919-1GEA
68.3
EUR
APOSOR17919-1G
APOSOR17919-5G
APOSOR17919-250MG
4-{[(2-Amino-4-hydroxypteridin-6-yl)methyl]amino}benzoic acid 95%
4-{[(2-Amino-4-hydroxypteridin-6-yl)methyl]amino}benzoic acid
Formel:
C₁₄H₁₂N₆O₃ Molecular Weight: 312.283 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075823 CAS-Nummer: 119-24-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...