5-Iodoindan-1-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,3-Dihydro-5-iodo-1H-inden-1-one, 5-Iodo-2,3-dihydro-1H-inden-1-one, 2,3-Dihydro-5-iodo-1-oxo-1H-indene
OR17925-1G
OR17925-250MG
APOSOR17925-1GEA
184
EUR
APOSOR17925-1G
APOSOR17925-250MG
5-Iodoindan-1-one
5-Iodoindan-1-one
Formel:
C₉H₇IO Molecular Weight: 258,056 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09744086 CAS-Nummer: 511533-38-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...