Methyl 3-bromoisoxazole-5-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromo-5-(methoxycarbonyl)isoxazole, Methyl 3-bromo-1,2-oxazole-5-carboxylate
OR17926-250MG
OR17926-500MG
OR17926-1G
APOSOR17926-250MGEA
168
EUR
APOSOR17926-250MG
APOSOR17926-500MG
APOSOR17926-1G
Methyl 3-bromoisoxazole-5-carboxylate
Methyl 3-bromoisoxazole-5-carboxylate
Formel:
C₅H₄BrNO₃ Molecular Weight: 205.994 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12022344 CAS-Nummer: 272773-11-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...