4-Methyl-3-nitrophenylacetic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(Carboxymethyl)-2-methylnitrobenzene, 4-(Carboxymethyl)-2-nitrotoluene, 2-(4-Methyl-3-nitrophenyl)acetic acid
OR18015-1G
OR18015-250MG
APOSOR18015-1GEA
53.8
EUR
APOSOR18015-1G
APOSOR18015-250MG
4-Methyl-3-nitrophenylacetic acid
4-Methyl-3-nitrophenylacetic acid
Formel:
C₉H₉NO₄ Molecular Weight: 195.172 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09832994 CAS-Nummer: 54941-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...