5-Bromo-4-methyl-1,3-thiazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-4-methylthiazole

OR18109-1G OR18109-250MG OR18109-5G
APOSOR18109-1GEA 58 EUR
APOSOR18109-1G APOSOR18109-250MG APOSOR18109-5G
5-Bromo-4-methyl-1,3-thiazole
5-Bromo-4-methyl-1,3-thiazole
Formel: C₄H₄BrNS
Molecular Weight: 178.05 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12026322
CAS-Nummer: 111600-83-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR