Methyl 7-aminothieno[2,3-b]pyrazine-6-carboxylate

Lieferant: Apollo Scientific
Achtung

OR183420-1G OR183420-5G
APOSOR183420-1GEA 87 EUR
APOSOR183420-1G APOSOR183420-5G
Methyl 7-aminothieno[2,3-b]pyrazine-6-carboxylate
Methyl 7-aminothieno[2,3-b]pyrazine-6-carboxylate
Formel: C₈H₇N₃O₂S
Molecular Weight: 209,225 g/mol
Schmelzpunkt: 130…133 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00204059
CAS-Nummer: 244014-85-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR