Methyl 3-amino-7-chlorobenzothiophene-2-carboxylate
Lieferant: Apollo Scientific
Synonyme:
Methyl 3-amino-7-chlorobenzo[b]thiophene-2-carboxylate
OR183426-1G
OR183426-250MG
APOSOR183426-1GEA
400
EUR
APOSOR183426-1G
APOSOR183426-250MG
Methyl 3-amino-7-chlorobenzothiophene-2-carboxylate
Methyl 3-amino-7-chlorobenzothiophene-2-carboxylate
Formel:
C₁₀H₈ClNO₂S Molecular Weight: 241,694 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD25960235 CAS-Nummer: 181283-33-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...