Methyl 3-amino-6-iodobenzothiophene-2-carboxylate
Lieferant: Apollo Scientific
OR183429-1G
OR183429-250MG
APOSOR183429-1GEA
400
EUR
APOSOR183429-1G
APOSOR183429-250MG
Methyl 3-amino-6-iodobenzothiophene-2-carboxylate
Methyl 3-amino-6-iodobenzothiophene-2-carboxylate
Formel:
C₁₀H₈INO₂S Molecular Weight: 333,145 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29163658 CAS-Nummer: 2089966-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...