4-(Thiomorpholin-4-yl)benzene-1,2-diamine

Lieferant: Apollo Scientific

OR183528-1G OR183528-250MG
APOSOR183528-1GEA 654 EUR
APOSOR183528-1G APOSOR183528-250MG
4-(Thiomorpholin-4-yl)benzene-1,2-diamine
4-(Thiomorpholin-4-yl)benzene-1,2-diamine
Formel: C₁₀H₁₅N₃S
Molecular Weight: 209,311 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1696692-81-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR