Furan-2-aldoxime
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Furan-2-carboxaldehyde oxime, 2-Furaldehyde oxime
OR18402-1G
OR18402-5G
OR18402-250MG
APOSOR18402-1GEA
36.4
EUR
APOSOR18402-1G
APOSOR18402-5G
APOSOR18402-250MG
Furan-2-aldoxime
Furan-2-aldoxime
Formel:
C₅H₅NO₂ Molecular Weight: 111,099 g/mol Schmelzpunkt: 78…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08445568 CAS-Nummer: 1121-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...