trans-4-(Dimethylamino)-4'-nitrochalcone

Lieferant: Apollo Scientific
Achtung

Synonyme: (2E)-3-[4-(Dimethylamino)phenyl]-1-(4-nitrophenyl)prop-2-en-1-one

OR1848-1G OR1848-250MG
APOSOR1848-1GEA 220 EUR
APOSOR1848-1G APOSOR1848-250MG
trans-4-(Dimethylamino)-4'-nitrochalcone
trans-4-(Dimethylamino)-4'-nitrochalcone
Formel: C₁₇H₁₆N₂O₃
Molecular Weight: 296,321 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01232245
CAS-Nummer: 20431-58-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR