3-Bromo-2-methoxybenzyl bromide

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Bromo-6-(bromomethyl)phenyl methyl ether, 1-Bromo-3-(bromomethyl)-2-methoxybenzene

OR18742-1G OR18742-250MG
APOSOR18742-1GEA 352 EUR
APOSOR18742-1G APOSOR18742-250MG
3-Bromo-2-methoxybenzyl bromide
3-Bromo-2-methoxybenzyl bromide
Formel: C₈H₈Br₂O
Molecular Weight: 279,957 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14656794
CAS-Nummer: 1177558-47-2
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR