3-(Piperazin-1-yl)benzoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(3-Carboxyphenyl)piperazine
OR1886-1G
OR1886-5G
APOSOR1886-1GEA
219
EUR
APOSOR1886-1G
APOSOR1886-5G
3-(Piperazin-1-yl)benzoic acid 97%
3-(Piperazin-1-yl)benzoic acid
Formel:
C₁₁H₁₄N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03444416 CAS-Nummer: 446831-28-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...