Methyl 4-(methoxymethyl)phenylacetate

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Methoxymethyl)phenylacetic acid methyl ester

OR19481-1G OR19481-5G
APOSOR19481-1GEA 145 EUR
APOSOR19481-1G APOSOR19481-5G
Methyl 4-(methoxymethyl)phenylacetate
Methyl 4-(methoxymethyl)phenylacetate
Formel: C₁₁H₁₄O₃
Molecular Weight: 194.227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12674804
CAS-Nummer: 115414-80-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR