2,6-Dibromobenzo[b]furan-3(2H)-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,6-Dibromo-1-benzofuran-3(2H)-one, 2,6-Dibromo-2,3-dihydro-3-oxobenzo[b]furan
OR19505-1G
OR19505-250MG
APOSOR19505-1GEA
560
EUR
APOSOR19505-1G
APOSOR19505-250MG
2,6-Dibromobenzo[b]furan-3(2H)-one
2,6-Dibromobenzo[b]furan-3(2H)-one
Formel:
C₈H₄Br₂O₂ Molecular Weight: 291,924 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD28096609 CAS-Nummer: 1823370-21-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...