Methyl 5-bromo-2-methoxy-3-nitrobenzoate

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2-(methoxycarbonyl)-6-nitroanisole, Methyl 5-bromo-3-nitro-o-anisate

OR19526-1G OR19526-250MG OR19526-5G
APOSOR19526-1GEA 406 EUR
APOSOR19526-1G APOSOR19526-250MG APOSOR19526-5G
Methyl 5-bromo-2-methoxy-3-nitrobenzoate
Methyl 5-bromo-2-methoxy-3-nitrobenzoate
Formel: C₉H₈BrNO₅
Molecular Weight: 290.068 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21340336
CAS-Nummer: 67657-90-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR