Methyl 3-amino-5-bromo-2-methoxybenzoate

Lieferant: Apollo Scientific
Achtung

OR19532-1G OR19532-250MG
APOSOR19532-1GEA 290 EUR
APOSOR19532-1G APOSOR19532-250MG
Methyl 3-amino-5-bromo-2-methoxybenzoate
Methyl 3-amino-5-bromo-2-methoxybenzoate
Formel: C₉H₁₀BrNO₃
Molecular Weight: 260,085 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23701735
CAS-Nummer: 926069-76-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR