Methyl 2-oxo-4-phenylbutanoate

Lieferant: Apollo Scientific

Synonyme: 2-Oxo-4-phenylbutyric acid methyl ester

OR1986-500MG OR1986-100MG
APOSOR1986-500MGEA 139 EUR
APOSOR1986-500MG APOSOR1986-100MG
Methyl 2-oxo-4-phenylbutanoate
Methyl 2-oxo-4-phenylbutanoate
Formel: C₁₁H₁₂O₃
Molecular Weight: 192,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08056218
CAS-Nummer: 83402-87-3
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR