3,5-Dichloropyridine-2-carbonyl chloride

Lieferant: Apollo Scientific

OR200113-1G OR200113-50MG OR200113-250MG OR200113-500MG
APOSOR200113-1GEA 432 EUR
APOSOR200113-1G APOSOR200113-50MG APOSOR200113-250MG APOSOR200113-500MG
3,5-Dichloropyridine-2-carbonyl chloride
3,5-Dichloropyridine-2-carbonyl chloride
Formel: C₆H₂Cl₃NO
Molecular Weight: 210,445 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18909534
CAS-Nummer: 70151-22-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR