2-Hydroxy-3-methoxybenzoyl chloride 95%
Lieferant: Apollo Scientific
OR200132-1G
OR200132-250MG
OR200132-500MG
APOSOR200132-1GEA
654
EUR
APOSOR200132-1G
APOSOR200132-250MG
APOSOR200132-500MG
2-Hydroxy-3-methoxybenzoyl chloride 95%
2-Hydroxy-3-methoxybenzoyl chloride
Formel:
C₈H₇ClO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11169659 CAS-Nummer: 82944-14-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...