2-(1H-Imidazol-1-yl)pyridine-4-carbonitrile
Lieferant: Apollo Scientific
OR200148-1G
OR200148-250MG
OR200148-500MG
APOSOR200148-1GEA
325
EUR
APOSOR200148-1G
APOSOR200148-250MG
APOSOR200148-500MG
2-(1H-Imidazol-1-yl)pyridine-4-carbonitrile
2-(1H-Imidazol-1-yl)pyridine-4-carbonitrile
Formel:
C₉H₆N₄ Molecular Weight: 170,171 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09929166 CAS-Nummer: 158020-84-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...