2-Amino-3-nitrobenzoyl chloride

Lieferant: Apollo Scientific

OR200152-1G OR200152-250MG OR200152-500MG OR200152-2.5G
APOSOR200152-1GEA 519 EUR
APOSOR200152-1G APOSOR200152-250MG APOSOR200152-500MG APOSOR200152-2.5G
2-Amino-3-nitrobenzoyl chloride
2-Amino-3-nitrobenzoyl chloride
Formel: C₇H₅ClN₂O₃
Molecular Weight: 200,579 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18389921
CAS-Nummer: 131645-73-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR