2-Pyrimidinecarbonyl chloride

Lieferant: Apollo Scientific
Gefahr

Synonyme: Pyrimidine-2-carbonyl chloride

OR200153-1G OR200153-250MG
APOSOR200153-1GEA 0 EUR
APOSOR200153-1G APOSOR200153-250MG
2-Pyrimidinecarbonyl chloride
2-Pyrimidinecarbonyl chloride
Formel: C₅H₃ClN₂O
Molecular Weight: 142.543 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18816666
CAS-Nummer: 220769-83-5
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR