3-(Ethoxycarbonyl)bicyclo[1.1.1]pentane-1-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR20017-1G OR20017-250MG OR20017-5G
APOSOR20017-1GEA 1350 EUR
APOSOR20017-1G APOSOR20017-250MG APOSOR20017-5G
3-(Ethoxycarbonyl)bicyclo[1.1.1]pentane-1-carboxylic acid
3-(Ethoxycarbonyl)bicyclo[1.1.1]pentane-1-carboxylic acid
Formel: C₉H₁₂O₄
Molecular Weight: 184,189 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD26406271
CAS-Nummer: 1823373-90-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR