2-(3-Bromophenoxy)pyridine 97+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-3-[(pyridin-2-yl)oxy]benzene, 3-Bromophenyl pyridin-2-yl ether
OR2051-1G
OR2051-250MG
APOSOR2051-1GEA
195
EUR
APOSOR2051-1G
APOSOR2051-250MG
2-(3-Bromophenoxy)pyridine 97+%
2-(3-Bromophenoxy)pyridine
Formel:
C₁₁H₈BrNO Molecular Weight: 250,091 g/mol Schmelzpunkt: 63…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08741430 CAS-Nummer: 92545-83-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...