{3-[(1H-Pyrazol-1-yl)methyl]phenyl}methylamine 90%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-[(1H-Pyrazol-1-yl)methyl]benzylamine, (3-((1H-Pyrazol-1-yl)methyl)phenyl)methanamine, 1-[3-(Aminomethyl)benzyl]-1H-pyrazole
OR2065-1G
OR2065-250MG
APOSOR2065-1GEA
932
EUR
APOSOR2065-1G
APOSOR2065-250MG
{3-[(1H-Pyrazol-1-yl)methyl]phenyl}methylamine 90%
{3-[(1H-Pyrazol-1-yl)methyl]phenyl}methylamine
Formel:
C₁₁H₁₃N₃ Molecular Weight: 187.241 g/mol Siedepunkt: 121…124 °C (0.3 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD08572164 CAS-Nummer: 562803-76-3 UN: 2735 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...