3-[(Pyrrolidin-1-yl)methyl]benzeneboronic acid, pinacol ester 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)pyrrolidine, 1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]pyrrolidine, 3-(Pyrrolidinomethyl)phenylboronic acid, pinacol ester

OR2072-1G OR2072-250MG
APOSOR2072-1GEA 139 EUR
APOSOR2072-1G APOSOR2072-250MG
3-[(Pyrrolidin-1-yl)methyl]benzeneboronic acid, pinacol ester 97%
3-[(Pyrrolidin-1-yl)methyl]benzeneboronic acid, pinacol ester
Formel: C₁₇H₂₆BNO₂
Molecular Weight: 287.205 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08741425
CAS-Nummer: 884507-45-3
UN: 2922
ADR: 8+6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR