4-(3-Methyl-1,2,4-oxadiazol-5-yl)benzoic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR2078-1G
APOSOR2078-1GEA
0
EUR
APOSOR2078-1G
4-(3-Methyl-1,2,4-oxadiazol-5-yl)benzoic acid 95%
4-(3-Methyl-1,2,4-oxadiazol-5-yl)benzoic acid
Formel:
C₁₀H₈N₂O₃ Molecular Weight: 204,182 g/mol Schmelzpunkt: 267…269 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08741427 CAS-Nummer: 851048-56-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...