6-Phenylnicotinonitrile 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Phenylpyridine-3-carbonitrile, 3-Cyano-6-phenylpyridine
OR2090-1G
OR2090-250MG
OR2090-100MG
APOSOR2090-1GEA
132
EUR
APOSOR2090-1G
APOSOR2090-250MG
APOSOR2090-100MG
6-Phenylnicotinonitrile 97%
6-Phenylnicotinonitrile
Formel:
C₁₂H₈N₂ Molecular Weight: 180,205 g/mol Schmelzpunkt: 88…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06637420 CAS-Nummer: 39065-54-8 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...