1-(2-Chlorobenzyl)homopiperazine

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(2-Chlorobenzyl)-1,4-diazepane

OR21021-1G OR21021-250MG
APOSOR21021-1GEA 0 EUR
APOSOR21021-1G APOSOR21021-250MG
1-(2-Chlorobenzyl)homopiperazine
1-(2-Chlorobenzyl)homopiperazine
Formel: C₁₂H₁₇ClN₂
Molecular Weight: 224,73 g/mol
Schmelzpunkt: 54…56 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01312090
CAS-Nummer: 76141-88-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR