2-Amino-5-nitrobenzamide

Lieferant: Apollo Scientific

OR21159-1G
APOSOR21159-1GEA 219 EUR
APOSOR21159-1G
2-Amino-5-nitrobenzamide
2-Amino-5-nitrobenzamide
Formel: C₇H₇N₃O₃
Molecular Weight: 181,149 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00090235
CAS-Nummer: 16313-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR