N'-(6-Chloropyridazin-3-yl)-N,N-dimethyliminoformamide

Lieferant: Apollo Scientific

OR21378-1G OR21378-250MG OR21378-500MG
APOSOR21378-1GEA 223 EUR
APOSOR21378-1G APOSOR21378-250MG APOSOR21378-500MG
N'-(6-Chloropyridazin-3-yl)-N,N-dimethyliminoformamide
N'-(6-Chloropyridazin-3-yl)-N,N-dimethyliminoformamide
Formel: C₇H₉ClN₄
Molecular Weight: 184,626 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00112306
CAS-Nummer: 35053-55-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR