1-Acetamido-3-aminonaphthalene

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(Acetylamino)-3-aminonaphthalene, N-(3-Aminonaphth-1-yl)acetamide

OR2139-250MG OR2139-500MG OR2139-50MG
APOSOR2139-250MGEA 522 EUR
APOSOR2139-250MG APOSOR2139-500MG APOSOR2139-50MG
1-Acetamido-3-aminonaphthalene
1-Acetamido-3-aminonaphthalene
Formel: C₁₂H₁₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09263886
CAS-Nummer: 721970-24-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR