2-{4-[(4-Methylbenzyl)oxy]phenyl}acetonitrile

Lieferant: Apollo Scientific

OR21494-10G OR21494-1G OR21494-5G
APOSOR21494-10GEA 346 EUR
APOSOR21494-10G APOSOR21494-1G APOSOR21494-5G
2-{4-[(4-Methylbenzyl)oxy]phenyl}acetonitrile
2-{4-[(4-Methylbenzyl)oxy]phenyl}acetonitrile
Formel: C₁₆H₁₅NO
Molecular Weight: 237,296 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00067740
CAS-Nummer: 175135-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR