2-[(5-Chloro-3-pyridyl)oxy]-3-nitropyridine

Lieferant: Apollo Scientific

OR21558-1G OR21558-5G OR21558-10G
APOSOR21558-1GEA 79.9 EUR
APOSOR21558-1G APOSOR21558-5G APOSOR21558-10G
2-[(5-Chloro-3-pyridyl)oxy]-3-nitropyridine
2-[(5-Chloro-3-pyridyl)oxy]-3-nitropyridine
Formel: C₁₀H₆ClN₃O₃
Molecular Weight: 251.626 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00067786
CAS-Nummer: 175135-51-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR