Methyl 4,5-dihydro-4-(2-methoxy-2-oxoethyl)-2-methyl-5-oxo-1H-pyrrole-3-carboxylate

Lieferant: Apollo Scientific
Achtung

OR21804-1G OR21804-5G OR21804-10G
APOSOR21804-1GEA 21.9 EUR
APOSOR21804-1G APOSOR21804-5G APOSOR21804-10G
Methyl 4,5-dihydro-4-(2-methoxy-2-oxoethyl)-2-methyl-5-oxo-1H-pyrrole-3-carboxylate
Methyl 4,5-dihydro-4-(2-methoxy-2-oxoethyl)-2-methyl-5-oxo-1H-pyrrole-3-carboxylate
Formel: C₁₀H₁₃NO₅
Molecular Weight: 227,214 g/mol
Schmelzpunkt: 93…94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173858
CAS-Nummer: 77978-74-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR