Ethyl 3,5-dimethyl-4-ethyl-1H-pyrrole-2-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4-Dimethyl-5-(ethoxycarbonyl)-3-ethyl-1H-pyrrole, 3,5-Dimethyl-2-(ethoxycarbonyl)-4-ethyl-1H-pyrrole

OR21807-1G OR21807-250MG
APOSOR21807-1GEA 140 EUR
APOSOR21807-1G APOSOR21807-250MG
Ethyl 3,5-dimethyl-4-ethyl-1H-pyrrole-2-carboxylate
Ethyl 3,5-dimethyl-4-ethyl-1H-pyrrole-2-carboxylate
Formel: C₁₁H₁₇NO₂
Molecular Weight: 195.258 g/mol
Schmelzpunkt: 87…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00030376
CAS-Nummer: 2199-47-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR